Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108667.1 | NP_001102137.1 | 7 - 8 | 1236 | 154 | |
XM_006238166.2 | XP_006238228.1 | 9 - 10 | 1343 | 154 | ||
XM_006238168.2 | XP_006238230.1 | 8 - 9 | 1270 | 154 | ||
XM_006238169.1 | XP_006238231.1 | 7 - 8 | 1153 | 154 | ||
XM_008763712.1 | XP_008761934.1 | 8 - 9 | 1363 | 154 | ||
XM_008763713.1 | XP_008761935.1 | 9 - 10 | 1337 | 154 | ||
XM_008763715.1 | XP_008761937.1 | 8 - 9 | 1270 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |