Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024764.1 | NP_001019935.1 | 4 - 5 | 542 | 94 | |
XM_006246381.2 | XP_006246443.1 | 4 - 5 | 585 | 94 | ||
XM_008767685.1 | XP_008765907.1 | 5 - 6 | 522 | 94 | ||
GenBank mRNA | BC097463.1 | - | 4 - 5 | 542 | 7457579 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |