Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105807.1 | NP_001099277.1 | 2 - 3 | 1065 | 81 | |
XM_008767959.1 | XP_008766181.1 | 2 - 3 | 11712 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |