Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105819.1 | NP_001099289.1 | 5 - 6 | 915 | 67 | |
XM_006246980.2 | XP_006247042.1 | 4 - 5 | 952 | 67 | ||
XM_006246981.1 | XP_006247043.1 | 5 - 6 | 1641 | 67 | ||
XM_006246982.2 | XP_006247044.1 | 5 - 6 | 977 | 67 | ||
XM_008767963.1 | XP_008766185.1 | 4 - 5 | 752 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |