Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gntl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015035.1 | NP_001015035.1 | 5 - 6 | 500 | 91 | |
XM_006247904.2 | XP_006247966.1 | 5 - 6 | 550 | 91 | ||
XM_008768503.1 | XP_008766725.1 | 5 - 6 | 550 | 91 | ||
XM_008768504.1 | XP_008766726.1 | 5 - 6 | 550 | 91 | ||
XM_008768505.1 | XP_008766727.1 | 5 - 6 | 550 | 91 | ||
XM_008768506.1 | XP_008766728.1 | 5 - 6 | 550 | 91 | ||
XM_008768507.1 | XP_008766729.1 | 5 - 6 | 550 | 91 | ||
XM_008768508.1 | XP_008766730.1 | 5 - 6 | 550 | 91 | ||
XM_008768509.1 | XP_008766731.1 | 5 - 6 | 550 | 91 | ||
XM_008768510.1 | XP_008766732.1 | 3 - 4 | 253 | 91 | ||
GenBank mRNA | AY634365.1 | - | 5 - 6 | 500 | 91 | |
BC111711.1 | - | 5 - 6 | 474 | 7383845 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |