Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108297.1 | NP_001101767.1 | 1 - 2 | 92 | 59 | |
GenBank mRNA | BC112385.1 | - | 1 - 2 | 76 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |