Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768165.1 | XP_008766387.1 | 17 - 18 | 3867 | 79 | |
XM_213427.7 | XP_213427.5 | 17 - 18 | 3867 | 79 | ||
GenBank mRNA | AF199337.1 | - | 17 - 18 | 2761 | 79 | |
AF199338.1 | - | 17 - 18 | 2761 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |