Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107055.1 | NP_001100525.1 | 2 - 3 | 391 | 89 | |
XM_006247529.2 | XP_006247591.1 | 2 - 3 | 1201 | 89 | ||
XM_006247530.2 | XP_006247592.1 | 3 - 4 | 1406 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Inventoried Probe spans exons |