Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106986.1 | NP_001100456.1 | 3 - 4 | 637 | 62 | |
XM_006245948.1 | XP_006246010.1 | 3 - 4 | 262 | 62 | ||
XM_006245949.1 | XP_006246011.1 | 2 - 3 | 649 | 62 | ||
XM_006245950.2 | XP_006246012.1 | 2 - 3 | 649 | 62 | ||
XM_008767566.1 | XP_008765788.1 | 2 - 3 | 649 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |