Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106981.1 | NP_001100451.1 | 2 - 3 | 629 | 95 | |
XM_006245870.2 | XP_006245932.1 | 2 - 3 | 629 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |