Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173374.1 | NP_001166845.1 | 5 - 6 | 752 | 86 | |
GenBank mRNA | BC158882.1 | - | 5 - 6 | 687 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |