Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105765.1 | NP_001099235.1 | 5 - 6 | 821 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |