Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172845.1 | NP_001166316.1 | 1 - 2 | 142 | 77 | |
GenBank mRNA | FQ221080.1 | - | 1 - 2 | 150 | 77 | |
FQ221398.1 | - | 1 - 2 | 150 | 77 | ||
FQ222010.1 | - | 1 - 2 | 153 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |