Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kremen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105767.1 | NP_001099237.1 | 6 - 7 | 781 | 67 | |
XM_008767550.1 | XP_008765772.1 | 5 - 6 | 1004 | 67 | ||
XM_008767551.1 | XP_008765773.1 | 5 - 6 | 1004 | 67 | ||
XM_008767552.1 | XP_008765774.1 | 5 - 6 | 1004 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |