Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080889.1 | NP_543165.1 | 5 - 6 | 769 | 76 | |
GenBank mRNA | AF410926.1 | - | 5 - 6 | 689 | 76 | |
AY325195.1 | - | 597 | 76 | |||
BC079343.1 | - | 5 - 6 | 769 | 7106866 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |