Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem41a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100805.1 | NP_001094275.1 | 1 - 2 | 177 | 92 | |
GenBank mRNA | BC161997.1 | - | 1 - 2 | 177 | 9035693 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |