Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142363.1 | NP_001135835.1 | 2 - 3 | 309 | 65 | |
XM_006248558.2 | XP_006248620.1 | 2 - 3 | 327 | 65 | ||
GenBank mRNA | CO401508.1 | - | 2 - 3 | 298 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |