Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106054.1 | NP_001099524.1 | 5 - 6 | 1899 | 123 | |
XM_008770864.1 | XP_008769086.1 | 5 - 6 | 2055 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |