Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012044.1 | NP_001012044.1 | 14 - 15 | 1673 | 80 | |
XM_006252318.2 | XP_006252380.1 | 14 - 15 | 1848 | 80 | ||
XM_006252319.2 | XP_006252381.1 | 16 - 17 | 2047 | 80 | ||
XM_006252320.2 | XP_006252382.1 | 16 - 17 | 2095 | 80 | ||
XM_006252322.2 | XP_006252384.1 | 14 - 15 | 3254 | 80 | ||
GenBank mRNA | BC083855.1 | - | 14 - 15 | 1673 | 7125327 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |