Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017505.1 | NP_001017505.1 | 6 - 7 | 964 | 80 | |
GenBank mRNA | BC087076.1 | - | 6 - 7 | 964 | 7134947 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |