Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053979.2 | NP_446431.1 | 4 - 5 | 524 | 93 | |
XM_008761696.1 | XP_008759918.1 | 4 - 5 | 596 | 93 | ||
GenBank mRNA | BC097294.1 | - | 4 - 5 | 524 | 7458249 | 93 |
X78604.1 | - | 4 - 5 | 373 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |