Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106937.1 | NP_001100407.1 | 5 - 6 | 668 | 160 | |
XM_006257159.2 | XP_006257221.1 | 6 - 7 | 937 | 160 | ||
XM_006257160.2 | XP_006257222.1 | 6 - 7 | 787 | 160 | ||
XM_006257161.2 | XP_006257223.1 | 6 - 7 | 934 | 160 | ||
XM_006257162.2 | XP_006257224.1 | 5 - 6 | 977 | 160 | ||
XM_008773339.1 | XP_008771561.1 | 5 - 6 | 652 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |