Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025288.1 | NP_001020459.1 | 1 - 2 | 217 | 71 | |
GenBank mRNA | BC097356.1 | - | 1 - 2 | 217 | 7442140 | 71 |
Gene Symbol: LOC100909986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003752209.3 | XP_003752257.1 | 2 - 3 | 462 | 71 | |
XM_006257731.2 | XP_006257793.1 | 2 - 3 | 462 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |