Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212490.2 | NP_997655.1 | 1 - 2 | 276 | 65 | |
GenBank mRNA | FQ212526.1 | - | 1 - 2 | 205 | 65 | |
FQ212959.1 | - | 1 - 2 | 238 | 65 | ||
FQ213756.1 | - | 1 - 2 | 129 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |