Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198744.2 | NP_942039.1 | 1 - 2 | 172 | 65 | |
XM_006235754.2 | XP_006235816.1 | 1 - 2 | 171 | 65 | ||
XM_006235757.2 | XP_006235819.1 | 1 - 2 | 191 | 65 | ||
XM_006235759.2 | XP_006235821.1 | 1 - 2 | 164 | 65 | ||
XM_006235760.2 | XP_006235822.1 | 1 - 2 | 186 | 65 | ||
XM_006235761.2 | XP_006235823.1 | 1 - 2 | 169 | 65 | ||
GenBank mRNA | BC059167.1 | - | 1 - 2 | 173 | 6890828 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |