Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108397.1 | NP_001101867.1 | 9 - 10 | 3517 | 102 | |
XM_008771124.1 | XP_008769346.1 | 8 - 9 | 3104 | 102 | ||
XM_008771125.1 | XP_008769347.1 | 9 - 10 | 3144 | 102 | ||
XM_008771126.1 | XP_008769348.1 | 9 - 10 | 3279 | 102 | ||
XM_008771127.1 | XP_008769349.1 | 8 - 9 | 3101 | 102 | ||
XM_008771128.1 | XP_008769350.1 | 9 - 10 | 3179 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |