Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011919.1 | NP_001011919.1 | 2 - 3 | 352 | 62 | |
GenBank mRNA | BC087083.1 | - | 2 - 3 | 352 | 7189218 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |