Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037778.1 | NP_001032867.1 | 4 - 5 | 406 | 117 | |
GenBank mRNA | BC098764.1 | - | 4 - 5 | 406 | 7371224 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |