Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017378.1 | NP_001017378.1 | 3 - 4 | 472 | 111 | |
GenBank mRNA | BC083891.1 | - | 3 - 4 | 472 | 7132551 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |