Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008890.1 | NP_001008890.1 | 1 - 2 | 93 | 59 | |
GenBank mRNA | BC157809.1 | - | 1 - 2 | 99 | 9033183 | 59 |
FQ222123.1 | - | 1 - 2 | 148 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |