Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748959.3 | XP_003749007.1 | 2 - 3 | 197 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |