Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005885.2 | NP_001005885.1 | 5 - 6 | 614 | 85 | |
XM_006229766.2 | XP_006229828.1 | 5 - 6 | 679 | 85 | ||
GenBank mRNA | AY325765.1 | - | 5 - 6 | 654 | 85 | |
AY380804.1 | - | 5 - 6 | 672 | 85 | ||
BC098002.1 | - | 5 - 6 | 614 | 7453525 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |