Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014244.1 | NP_001014266.1 | 5 - 6 | 503 | 93 | |
XM_008759774.1 | XP_008757996.1 | 3 - 4 | 1641 | 93 | ||
XM_008759775.1 | XP_008757997.1 | 4 - 5 | 1544 | 93 | ||
XM_008759776.1 | XP_008757998.1 | 3 - 4 | 1641 | 93 | ||
XM_008759777.1 | XP_008757999.1 | 3 - 4 | 1641 | 93 | ||
XM_008759778.1 | XP_008758000.1 | 4 - 5 | 959 | 93 | ||
XM_008759779.1 | XP_008758001.1 | 4 - 5 | 1116 | 93 | ||
XM_008759780.1 | XP_008758002.1 | 2 - 3 | 145 | 93 | ||
XM_008759781.1 | XP_008758003.1 | 5 - 6 | 440 | 93 | ||
XM_008759782.1 | XP_008758004.1 | 3 - 4 | 1641 | 93 | ||
GenBank mRNA | BC079235.1 | - | 5 - 6 | 503 | 7132617 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |