Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105712.1 | NP_001099182.1 | 3 - 4 | 1551 | 110 | |
XM_008762681.1 | XP_008760903.1 | 2 - 3 | 3244 | 110 | ||
XM_008762682.1 | XP_008760904.1 | 3 - 4 | 2027 | 110 | ||
XM_008762683.1 | XP_008760905.1 | 2 - 3 | 3244 | 110 | ||
GenBank mRNA | M92076.1 | - | 2 - 3 | 1469 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |