Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106421.1 | NP_001099891.1 | 4 - 5 | 536 | 111 | |
XM_006232142.1 | XP_006232204.1 | 4 - 5 | 800 | 111 | ||
GenBank mRNA | GQ499323.1 | - | 4 - 5 | 503 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |