Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106425.1 | NP_001099895.1 | 2 - 3 | 392 | 66 | |
XM_008760864.1 | XP_008759086.1 | 2 - 3 | 401 | 66 | ||
GenBank mRNA | BC158721.1 | - | 2 - 3 | 383 | 9090461 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |