Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Comtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107249.1 | NP_001100719.1 | 1 - 2 | 161 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |