Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107159.1 | NP_001100629.1 | 2 - 3 | 264 | 81 | |
XM_006240758.2 | XP_006240820.1 | 2 - 3 | 184 | 81 | ||
GenBank mRNA | FJ799755.1 | - | 2 - 3 | 264 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |