Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769053.1 | XP_008767275.1 | 12 - 13 | 1255 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |