Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109061.1 | NP_001102531.1 | 10 - 11 | 1045 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons |