Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108875.2 | NP_001102345.2 | 3 - 4 | 556 | 122 | |
XM_008771302.1 | XP_008769524.1 | 3 - 4 | 390 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |