Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012847.1 | NP_036979.1 | 6 - 7 | 838 | 74 | |
GenBank mRNA | BC085758.1 | - | 6 - 7 | 788 | 7189836 | 74 |
M81225.1 | - | 6 - 7 | 838 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |