Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014143.1 | NP_001014165.1 | 5 - 6 | 804 | 107 | |
GenBank mRNA | BC089828.1 | - | 5 - 6 | 804 | 7322740 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |