Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751621.3 | XP_003751669.1 | 11 - 12 | 1089 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |