Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013225.2 | NP_001013243.2 | 8 - 9 | 1348 | 58 | |
XM_006244543.2 | XP_006244605.1 | 8 - 9 | 1390 | 58 | ||
XM_006244544.2 | XP_006244606.1 | 8 - 9 | 1180 | 58 | ||
XM_006244545.2 | XP_006244607.1 | 7 - 8 | 3264 | 58 | ||
XM_006244546.2 | XP_006244608.1 | 8 - 9 | 1169 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |