Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007801.2 | NP_001007802.1 | 5 - 6 | 631 | 75 | |
GenBank mRNA | AY387096.1 | - | 5 - 6 | 526 | 75 | |
BC087102.1 | - | 5 - 6 | 631 | 7192818 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |