Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015011.1 | NP_001015011.1 | 4 - 5 | 434 | 84 | |
GenBank mRNA | BC091568.1 | - | 4 - 5 | 434 | 7443410 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |