Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277272.1 | NP_001264201.1 | 689 | 72 | ||
XM_006229555.2 | XP_006229617.1 | 1178 | 72 | |||
GenBank mRNA | BC103640.1 | - | 580 | 7134953 | 72 | |
BC158688.1 | - | 621 | 8375819 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |