Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231862.2 | XP_006231924.1 | 12 - 13 | 3972 | 120 | |
XM_006231863.2 | XP_006231925.1 | 12 - 13 | 3776 | 120 | ||
XM_008760695.1 | XP_008758917.1 | 16 - 17 | 4323 | 120 | ||
XM_008760696.1 | XP_008758918.1 | 15 - 16 | 4218 | 120 | ||
XM_008760697.1 | XP_008758919.1 | 15 - 16 | 4204 | 120 | ||
XM_008760698.1 | XP_008758920.1 | 11 - 12 | 3638 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Probe spans exons |