Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231862.2 | XP_006231924.1 | 14 - 15 | 4221 | 133 | |
XM_006231863.2 | XP_006231925.1 | 14 - 15 | 4025 | 133 | ||
XM_008760695.1 | XP_008758917.1 | 18 - 19 | 4572 | 133 | ||
XM_008760696.1 | XP_008758918.1 | 17 - 18 | 4467 | 133 | ||
XM_008760697.1 | XP_008758919.1 | 17 - 18 | 4453 | 133 | ||
XM_008760698.1 | XP_008758920.1 | 13 - 14 | 3887 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Probe spans exons |